missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL5 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82063F
This item is not returnable.
View return policy
Description
YPEL5 Polyclonal antibody specifically detects YPEL5 in Human, Mouse samples. It is validated for ELISA, Immunofluorescence
Specifications
| YPEL5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunofluorescence | |
| FITC | |
| CGI-127, protein yippee-like 5, yippee-like 5 (Drosophila) | |
| YPEL5 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human YPEL5. The immunogen is located within the first 50 amino acids of YPEL5. Amino Acid Squence: AAANCDTILTNRSELI | |
| 0.1 mL | |
| Cell Biology | |
| 51646 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction