missing translation for 'onlineSavingsMsg'
Learn More

YPEL4 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18425617 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425617 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425617 Supplier Novus Biologicals Supplier No. NBP281957C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

YPEL4 Polyclonal antibody specifically detects YPEL4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen YPEL4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias FLJ30213, MGC102723, MGC138324, protein yippee-like 4, yippee-like 4 (Drosophila)
Host Species Rabbit
Immunogen YPEL4 antibody was raised against a 12 amino acid synthetic peptide near the amino terminus of human YPEL4. The immunogen is located within the first 50 amino acids of YPEL4. Amino Acid Squence: LPTKTFRSYLPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 219539
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.