missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ YPD Broth (Powder/Molecular Genetics)
£23.72 - £876.00
Specifications
Content And Storage | 4°C |
---|---|
Form | Powder |
Grade | Molecular genetics |
Media Type | YPD Broth |
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10306513
|
Fisher BioReagents™
BP2469-500 |
500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10205163
|
Fisher BioReagents™
BP2469-100 |
100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10797535
|
Fisher BioReagents™
BP2469-10 |
10,000 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10215163
|
Fisher BioReagents™
BP2469-2 |
2000 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
4°C | |
Molecular genetics | |
Microbiology Media |
Powder | |
YPD Broth |