missing translation for 'onlineSavingsMsg'
Learn More

Yohimbine Hydrochloride, MedChemExpress

Product Code. 16472499
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472499 Supplier Medchem Express Supplier No. HYN0127
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Yohimbine Hydrochloride is an alpha 2-adrenoreceptor antagonist, blocking the pre- and postsynaptic alpha-2 adrenoreceptors and causing an increased release of noradrenaline and dopamine.

Yohimbine Hydrochloride is an alpha 2-adrenoreceptor antagonist, blocking the pre- and postsynaptic alpha-2 adrenoreceptors and causing an increased release of noradrenaline and dopamine.

Specifications

Chemical Name or Material Yohimbine Hydrochloride
CAS 65-19-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H27ClN2O3
Quantity 1 g
Solubility Information DMSO : 33.33 mg/mL (85.26 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (8.52 mM; Need ultrasonic)
SMILES [H][C@]12C(NC3=C4C=CC=C3)=C4CCN1C[C@@]5(CC[C@H](O)[C@H](C(OC)=O)[C@]5(C2)[H])[H].[H]Cl
Molecular Weight (g/mol) 390.9
Formula Weight 390.9
Percent Purity 99.53%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.