missing translation for 'onlineSavingsMsg'
Learn More

Yoda 1, TRC

Product Code. 30456560
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456560 10 mg 10mg
30453695 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456560 Supplier Toronto Research Chemicals Supplier No. TRCY10078510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448947-81-7
Molecular Formula C13H8Cl2N4S2
Molecular Weight (g/mol) 355.27
Synonym 2-[5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]pyrazine, [5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]-pyrazine
IUPAC Name 2-[(2,6-dichlorophenyl)methylsulfanyl]-5-pyrazin-2-yl-1,3,4-thiadiazole
SMILES Clc1cccc(Cl)c1CSc2nnc(s2)c3cnccn3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H8Cl2N4S2/c14-9-2-1-3-10(15)8(9)7-20-13-19-18-12(21-13)11-6-16-4-5-17-11/h1-6H,7H2
Formula Weight 353.9567
Chemical Name or Material Yoda 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.