missing translation for 'onlineSavingsMsg'
Learn More

Yoda 1, MedChemExpress

Product Code. p-200100531
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490078

Brand: Medchem Express HY18723

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Yoda 1 is a potent and selective Piezo1 agonist. Yoda 1 activates purified Piezo1 channels. Yoda1 induces activation of both Akt and ERK1/2 in endothelial cells (ECs), which is not dependent on Piezo1. Yoda 1 potently inhibits macropinocytosis induced by epidermal growth factor (EGF). Yoda 1 enhances Ca2+ influx followed by activation of the calcium-activated potassium channel KCa3.1 and inhibition of Rac1 activation.

Yoda 1 is a potent and selective Piezo1 agonist. Yoda 1 activates purified Piezo1 channels. Yoda1 induces activation of both Akt and ERK1/2 in endothelial cells (ECs), which is not dependent on Piezo1. Yoda 1 potently inhibits macropinocytosis induced by epidermal growth factor (EGF). Yoda 1 enhances Ca2+ influx followed by activation of the calcium-activated potassium channel KCa3.1 and inhibition of Rac1 activation.

Specifications

Chemical Name or Material Yoda 1
Physical Form Powder
CAS 448947-81-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H8Cl2N4S2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 15.62 mg/mL (43.97 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=C(CSC2=NN=C(C3=NC=CN=C3)S2)C(Cl)=CC=C1
Molecular Weight (g/mol) 355.27
Formula Weight 355.27
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.