missing translation for 'onlineSavingsMsg'
Learn More

Yoda 1, MedChemExpress

Product Code. 16490078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490078 5 mg 5mg
16477608 10 mg 10mg
16453627 50 mg 50mg
16437526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490078 Supplier Medchem Express Supplier No. HY18723
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Yoda 1 is a potent and selective Piezo1 agonist. Yoda 1 activates purified Piezo1 channels. Yoda1 induces activation of both Akt and ERK1/2 in endothelial cells (ECs), which is not dependent on Piezo1. Yoda 1 potently inhibits macropinocytosis induced by epidermal growth factor (EGF). Yoda 1 enhances Ca2+ influx followed by activation of the calcium-activated potassium channel KCa3.1 and inhibition of Rac1 activation.

Yoda 1 is a potent and selective Piezo1 agonist. Yoda 1 activates purified Piezo1 channels. Yoda1 induces activation of both Akt and ERK1/2 in endothelial cells (ECs), which is not dependent on Piezo1. Yoda 1 potently inhibits macropinocytosis induced by epidermal growth factor (EGF). Yoda 1 enhances Ca2+ influx followed by activation of the calcium-activated potassium channel KCa3.1 and inhibition of Rac1 activation.

Specifications

Chemical Name or Material Yoda 1
Physical Form Powder
CAS 448947-81-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H8Cl2N4S2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 15.62 mg/mL (43.97 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=C(CSC2=NN=C(C3=NC=CN=C3)S2)C(Cl)=CC=C1
Molecular Weight (g/mol) 355.27
Formula Weight 355.27
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.