missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Yoda 1

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Description
Yoda 1 is a selective activator of mouse and human mechanosensitive channel piezo1. In HEK cells, Yoda 1 slows the inactivation phase of transient currents, sensitizes Piezo1 to activation by pressure, and partially activates channels in the absence of external pressure. In MDCK cells, Yoda 1 induces Ca2+-dependent chromatin hypercondensation and alters Ca2+-dependent myosin contractility, leading to nuclear shrinkage in cells. In red blood cells from sickle cell anemia (SCA) patients, Yoda 1 increases intracellular Ca2+ and phosphatidylserine exposure, which leads to KCa3.1 channel mediated Ca2+ influx and K+ and water efflux, causing shrinkage of the cells.
Specifications
Specifications
| Chemical Name or Material | 2-[5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]pyrazine |
| CAS | 448947-81-7 |
| Quantity | 10 mg |
| Target | Other Calcium Channel Activators |
| Molecular Formula | C13H8Cl2N4S2 |
| Formula Weight | 355.27 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction