missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Yoda 1
GREENER_CHOICE

Product Code. 18775866
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18775866

Brand: Tocris Bioscience™ 5586/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Piezo1 channel activator

Yoda 1 is a selective activator of mouse and human mechanosensitive channel piezo1. In HEK cells, Yoda 1 slows the inactivation phase of transient currents, sensitizes Piezo1 to activation by pressure, and partially activates channels in the absence of external pressure. In MDCK cells, Yoda 1 induces Ca2+-dependent chromatin hypercondensation and alters Ca2+-dependent myosin contractility, leading to nuclear shrinkage in cells. In red blood cells from sickle cell anemia (SCA) patients, Yoda 1 increases intracellular Ca2+ and phosphatidylserine exposure, which leads to KCa3.1 channel mediated Ca2+ influx and K+ and water efflux, causing shrinkage of the cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]pyrazine
CAS 448947-81-7
Quantity 10 mg
Target Other Calcium Channel Activators
Molecular Formula C13H8Cl2N4S2
Formula Weight 355.27
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.