missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ YNT 185

Description
YNT 185 is a potent and selective orexin OX2 receptor agonist (EC50 = 28 nM at human OX2 expressed in CHO cells). Displays approximately 100-fold selectivity for OX2 over OX1 (EC50 = 2.75 μM at human OX1 expressed in CHO cells). Depolarizes OX2-expressing histaminergic neurons in mouse brain slices. Increases wake time in wild type mice. Suppresses cataplexy-like symptoms in OX knockout mice.
Specifications
Specifications
| Chemical Name or Material | 3-[[[3-[[2-[[2-(Dimethylamino)benzoyl]amino]ethyl]amino]phenyl]amino]sulfonyl]-4-methoxy-N,N-dimethyl-[1,1-biphenyl]-3-carboxamide dihydrochloride |
| CAS | 1804978-82-2 |
| Quantity | 10 mg |
| Target | OX2 Receptor Agonists |
| Molecular Formula | C33H37N5O5S0.2 HCl |
| Formula Weight | 2778 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?