missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ YNB with Ammonium Sulfate (Powder)

Product Code. 11387809 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
227 g
Unit Size:
227g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387809 227 g 227g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11387809 Supplier MP Biomedicals™ Supplier No. 114027512
  • ONLINE EXCLUSIVE PRICE
    £189.00
    £159.60 / 227g
    Save £29.40
    15% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Causes vigorous growth of Saccharomyces cerevisiae. MP Biomedicals YNB with Ammonium Sulfate (Powder) is a well-defined composition of salts, vitamins and a nitrogen source. It is a component of synthetic defined yeast media.

Specifications

Form Powder
For Use With (Application) YNB w/ Ammonium Sulfate is a component of synthetic defined yeast media.
Quantity 227 g
compliance-icons
Product Identifier
  • YNB with Ammonium Sulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ammonium sulphate, potassium dihydrogenphosphate, calcium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.