missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YME1L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564299
This item is not returnable.
View return policy
Description
Immunogen sequence: AVDGKEMVTM KELEFSKDKI LMGPERRSVE IDNKNKTITA YHESGHAIIA YYTKDAMPIN KATIMPRGPT LGHVSLLPEN DRWNETRAQL LAQMDVS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.
Specifications
| YME1L1 | |
| Polyclonal | |
| Unconjugated | |
| YME1L1 | |
| ATP-dependent metalloprotease FtsH1; ATP-dependent metalloprotease FtsH1 homolog; ATP-dependent metalloprotease YME1L1; ATP-dependent metalloprotease YME1L1-like protein; ATP-dependent zinc metalloprotease YME1L1; ATP-dependent zinc metalloprotease YME1l1-like protein; FTSH; FTSH1; MEG4; meg-4; PAMP; Presenilin-associated metalloprotease; UNQ1868/PRO4304; YME1 like 1 ATPase; YME1L; YME1L1; YME1-like 1 (S. cerevisiae); YME1-like 1 ATPase; YME1-like protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10730 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96TA2 | |
| YME1L1 | |
| Recombinant protein corresponding to Human YME1L1. Recombinant protein control fragment (Product #RP-103788). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction