Learn More
YMC Group YMC-Triart Prep C18-S, semi-preparative HPLC-column (10.0 mm ID), 12 nm, S-7 µm, 150 x 10.0 mm
Brand:  YMC Group TAS12S07-1510WT
Encompass_Preferred
YMC-Triart Prep C18-S, semi-preparative HPLC-column (10.0 mm ID), 12 nm, S-7 µm, 150 x 10.0 mm
Product Code. 17299675
  • £1623.65 / Each
Only null left
Estimated Shipment: 07-01-2026