Learn More
YMC Group YMC-Triart C18, Classical Analytical HPLC Column (4.0 mm ID), 12 nm, S-3 µm, 125 x 4.0 mm
Brand:  YMC Group TA12S03-R504WT
Encompass_Preferred
YMC-Triart C18, Classical Analytical HPLC Column (4.0 mm ID), 12 nm, S-3 µm, 125 x 4.0 mm
Product Code. 17228985
  • £1014.66 / Each
Only null left
Estimated Shipment: 07-01-2026