Learn More
YMC Group YMC-Pack Pro C18 RS, Classical Analytical HPLC column (4.6 mm ID), 8 nm, S-3 µm, 250 x 4.6 mm
Brand:  YMC Group RS08S03-2546WT
Encompass_Preferred
YMC-Pack Pro C18 RS, Classical Analytical HPLC column (4.6 mm ID), 8 nm, S-3 µm, 250 x 4.6 mm
Product Code. 17137728
  • £1243.08 / Each