Learn More
YMC Group YMC-Pack ODS-A, Microbore HPLC column (3.0 mm ID), 20 nm, S-3 µm, 100 x 3.0 mm
Brand:  YMC Group AA20S03-1003WT
Encompass_Preferred
YMC-Pack ODS-A, Microbore HPLC column (3.0 mm ID), 20 nm, S-3 µm, 100 x 3.0 mm
Product Code. 17293935
  • £889.79 / Each
Only null left
Estimated Shipment: 05-01-2026