Learn More
YMC Group YMC-Pack ODS-A-HG Prep, semi-preparative HPLC column (20.0 mm ID), 12 nm, S-50 µm, 250 x 20.0 mm
Brand:  YMC Group AAG12S50-2520WT
Encompass_Preferred
YMC-Pack ODS-A-HG Prep, semi-preparative HPLC column (20.0 mm ID), 12 nm, S-50 µm, 250 x 20.0 mm
Product Code. 17214125
  • POA / Pack of 1