Learn More
YMC Group YMC-Pack NH2 (Amino), semi-preparative HPLC column 30 mm ID, 12 nm, S-5 µm, 150 x 30.0 mm
Brand:  YMC Group NH12S05-1530WT
Encompass_Preferred
YMC-Pack NH2 (Amino), semi-preparative HPLC column 30 mm ID, 12 nm, S-5 µm, 150 x 30.0 mm
Product Code. 17267555
  • £7278.12 / Each
Only null left
Estimated Shipment: 06-01-2026