Learn More
YMC Group YMC-Pack C4, narrowbore HPLC colum n (2.0 mm ID),12 nm , S-3 µm, 150 x 2.0 mm
Brand:  YMC Group BU12S03-1502WT
Encompass_Preferred
YMC-Pack C4, narrowbore HPLC colum n (2.0 mm ID),12 nm , S-3 µm, 150 x 2.0 mm
Product Code. 30223809
  • £1030.05 / Each
Only null left
Estimated Shipment: 07-01-2026