missing translation for 'onlineSavingsMsg'
Learn More

YM17E, MedChemExpress

Product Code. 16460819
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460819 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16460819 Supplier Medchem Express Supplier No. HY101627

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress YM17E is an inhibitor of acyl CoA:cholesterol acyltransferase (ACAT), with IC50 of 44 nM in rabbit liver microsomes in vitro.

YM17E is an inhibitor of acyl CoA:cholesterol acyltransferase (ACAT), with IC50 of 44 nM in rabbit liver microsomes in vitro.

Specifications

Chemical Name or Material YM17E
Physical Form Solid
CAS 124900-72-7
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C40H56N6O2
For Use With (Application) Metabolism-sugar/lipid metabolism
SMILES O=C(NC1=CC=C(N(C)C)C=C1)N(CC2=CC=CC(CN(C3CCCCCC3)C(NC4=CC=C(N(C)C)C=C4)=O)=C2)C5CCCCCC5
Molecular Weight (g/mol) 652.91
Formula Weight 652.91
Percent Purity 97.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.