missing translation for 'onlineSavingsMsg'
Learn More

YM-90709, MedChemExpress

Product Code. 16443917
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443917

Brand: Medchem Express HY19969

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YM-90709 is a novel IL-5 inhibitor which selectively blocks the binding of IL-5 to the IL-5 receptor (IL-5R).YM-90709 potently inhibits the binding of [125I]-IL-5 to IL-5R on human peripheral eosinophils and eosinophilic HL-60 clone 15 cells with IC50 values of 1.0 and 0.57 μM.

YM-90709 is a novel IL-5 inhibitor which selectively blocks the binding of IL-5 to the IL-5 receptor (IL-5R).YM-90709 potently inhibits the binding of [125I]-IL-5 to IL-5R on human peripheral eosinophils and eosinophilic HL-60 clone 15 cells with IC50 values of 1.0 and 0.57 μM.

Specifications

Chemical Name or Material YM-90709
CAS 163769-88-8
Color Orange
Purity Grade Notes Research
Molecular Formula C22H21N3O2
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (173.89 mM; Need ultrasonic)
SMILES CC(CC1=CC(OC)=C(OC)C=C12)(C)N3C2=CC4=NC5=CC=CC=C5N=C43
Molecular Weight (g/mol) 359.42
Formula Weight 359.42
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.