missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ YM 244769
GREENER_CHOICE

Product Code. 30001586
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001586

Brand: Tocris Bioscience™ 4544/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Na+/Ca2+ exchange inhibitor (reverse mode); neuroprotective

Inhibitor of the reverse mode of Na+/Ca2+ exchange (NCX). Potently inhibits Na+-dependent 45Ca2+ uptake (IC50 values are 18, 68 and 96 nM for CCL39 cells transfected with NCX3, NCX1 and NCX2 respectively). Displays selectivity for NCX over other Na+ and Ca2+ transporters. Protects against hypoxia/reoxygenation-induced cell damage in neuronal SH-SY5Y cells expressing NCX1 and NCX3.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1780390-65-9
Molecular Formula C26H23ClFN3O3

Specifications

Quantity 50 mg
Formula Weight 516.39
Purity >99%
Molecular Weight (g/mol) 479.936
Chemical Name or Material YM 244769
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.