missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ YM 244769

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Inhibitor of the reverse mode of Na+/Ca2+ exchange (NCX). Potently inhibits Na+-dependent 45Ca2+ uptake (IC50 values are 18, 68 and 96 nM for CCL39 cells transfected with NCX3, NCX1 and NCX2 respectively). Displays selectivity for NCX over other Na+ and Ca2+ transporters. Protects against hypoxia/reoxygenation-induced cell damage in neuronal SH-SY5Y cells expressing NCX1 and NCX3.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 516.39 |
| Purity | >99% |
| Molecular Weight (g/mol) | 479.936 |
| Chemical Name or Material | YM 244769 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction