missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YM-201636, MedChemExpress
Description
YM-201636 is a potent and selective PIKfyve inhibitor with an IC50 of 33 nM. YM-201636 also inhibits p110α with an IC50 of 3.3 μM. YM-201636 inhibits retroviral replication.
Specifications
Specifications
| Chemical Name or Material | YM-201636 |
| CAS | 371942-69-7 |
| Color | Yellowish Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C25H21N7O3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 47 mg/mL (100.54 mM) |
| SMILES | O=C(NC1=CC=CC(C2=NC(N3CCOCC3)=C4OC5=NC=CC=C5C4=N2)=C1)C6=CC=C(N=C6)N |
| Molecular Weight (g/mol) | 467.48 |
| Formula Weight | 467.48 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?