missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YL1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71870PE
This item is not returnable.
View return policy
Description
YL1 Polyclonal antibody specifically detects YL1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| YL1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| Protein YL-1, Swc2, TCFL1, Transcription factor-like 1, transcription factor-like 1 (YL-1), vacuolar protein sorting 72 (yeast), vacuolar protein sorting 72 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 72 homolog, YL-1, YL1CFL1 | |
| The immunogen for this product maps to a region between residue 50 and 100 of human Protein YL-1 using the numbering given in entry NP_005988.1 (GeneID 6944). | |
| 0.1 mL | |
| Autophagy | |
| 6944 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction