missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ YL1 Antibody (2G6), Novus Biologicals™

Product Code. 18343639 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343639 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343639 Supplier Novus Biologicals™ Supplier No. H00006944M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

YL1 Monoclonal antibody specifically detects YL1 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen YL1
Applications ELISA, Western Blot, Sandwich ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2G6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH03151
Gene Alias Protein YL-1, Swc2, TCFL1, Transcription factor-like 1, transcription factor-like 1 (YL-1), vacuolar protein sorting 72 (yeast), vacuolar protein sorting 72 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 72 homolog, YL-1, YL1CFL1
Host Species Mouse
Immunogen VPS72 (AAH03151, 1 a.a. ∽ 364 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSLAGGRAPRKTAGNRLSGLLEAEEEDEFYQTTYGGFTEESGDDEYQGDQSDTEDEVDSDFDIDEGDEPSSDGEAEEPRRKRRVVTKAYKEPLKSLRPRKVNTPAGSSQKAREEKALLPLELQDDGSDSRKSMRQSTAEHTRQTFLRVQERQGQSRRRKGPHCERPLTQEELLREAKITEELNLRSLETYERLEADKKKQVHKKRKCPGPIITYHSVTVPLVGEPGPKEENVDIEGLDPAPSVSALTPHAGTGPVNPPARCSRTFITFSDDATFEEWFPQGRPPKVPVREVCPVTHRPALYRDPVTDIPYATARAFKIIREAYKKYITAHGLPPTASALGPGPPPPEPLPGSGPRALRQKIVIK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 6944
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.