missing translation for 'onlineSavingsMsg'
Learn More

YKL-05-099, MedChemExpress

Product Code. 16486007
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486007 5 mg 5mg
16459826 50 mg 50mg
16453288 10 mg 10mg
16404546 100 mg 100mg
16400739 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16486007 Supplier Medchem Express Supplier No. HY101147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YKL-05-099 is a salt-inducible kinase (SIK) inhibitor. YKL-05-099 binds to SIK1 and SIK3 with IC50s of ∼10 and ∼30 nM, respectively. YKL-05-099 has slightly less potent SIK2-inhibitory (IC50=40 nM).

YKL-05-099 is a salt-inducible kinase (SIK) inhibitor. YKL-05-099 binds to SIK1 and SIK3 with IC50s of ∼10 and ∼30 nM, respectively. YKL-05-099 has slightly less potent SIK2-inhibitory (IC50=40 nM).

Specifications

Chemical Name or Material YKL-05-099
CAS 1936529-65-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C32H34ClN7O3
Quantity 5 mg
Solubility Information DMSO : ≥ 75 mg/mL (124.98 mM)
SMILES O=C1N(C2=NC=C(OC)C=C2)C3=NC(NC4=CC=C(C5CCN(C)CC5)C=C4OC)=NC=C3CN1C6=C(C)C=CC=C6Cl
Molecular Weight (g/mol) 600.11
Formula Weight 600.11
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.