missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ YK 4-279
GREENER_CHOICE

Product Code. 18065852
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065852 50 mg 50mg
18054152 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065852 Supplier Tocris Bioscience™ Supplier No. 4067/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of RNA helicase A (RHA)

Inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1. Inhibits Ewing's sarcoma family tumor (ESFT) cell growth; induces apoptosis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1037184-44-3
Molecular Formula C17H13Cl2NO4

Specifications

Quantity 50 mg
Formula Weight 366.2
Purity >99%
Molecular Weight (g/mol) 366.19
Chemical Name or Material YK 4-279
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.