missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YIPF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571287
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Human: 100%; Pig: 77%.
YIPF7 is a protein coding gene.
Specifications
| YIPF7 | |
| Polyclonal | |
| Unconjugated | |
| YIPF7 | |
| FinGER9; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 9; protein YIPF7; RGD1310766; Yip1 domain family member 7; Yip1 domain family, member 7; YIP1 family member 7; YIP1B; YIPF7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 285525 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N8F6 | |
| YIPF7 | |
| synthetic peptide directed towards the N-terminal region of human YIPF7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction