missing translation for 'onlineSavingsMsg'
Learn More

YIPF5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16870894 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16870894 20 μL 20µL
16860894 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16870894 Supplier Proteintech Supplier No. 129311AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Plays a role in transport between endoplasmic reticulum and Golgi.

Specifications

Antigen YIPF5
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene YIPF5
Gene Accession No. Q5XID0, Q969M3, Q9EQQ2
Gene Alias FinGER5, Protein YIPF5, SB140, SMAP 5, SMAP5, UNQ3123/PRO10275, Yip1 domain family, member 5, YIP1 family member 5, YIP1A, YIPF5, YPT interacting protein 1 A
Gene Symbols Yipf5
Host Species Rabbit
Immunogen YIPF5 Fusion Protein Ag3802
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361315, 67180, 81555
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.