missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YIPF5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12931-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a role in transport between endoplasmic reticulum and Golgi.Specifications
| YIPF5 | |
| Polyclonal | |
| Unconjugated | |
| YIPF5 | |
| FinGER5, Protein YIPF5, SB140, SMAP 5, SMAP5, UNQ3123/PRO10275, Yip1 domain family, member 5, YIP1 family member 5, YIP1A, YIPF5, YPT interacting protein 1 A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361315, 67180, 81555 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5XID0, Q969M3, Q9EQQ2 | |
| Yipf5 | |
| YIPF5 Fusion Protein Ag3802 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction