Learn More
Invitrogen™ YIPF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139922
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MAAVDDLQFE EFGNAATSLT ANPDATTVNI EDPGETPKHQ PGSPRGSGRE ED Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 73%.
YIPF1 is a multi-pass membrane protein. It belongs to the YIP1 family. The exact function of YIPF1 remains unknown.
Specifications
| YIPF1 | |
| Polyclonal | |
| Unconjugated | |
| YIPF1 | |
| C030002N13Rik; DJ167A19.1; FinGER1; protein YIPF1; Yip1 domain family member 1; Yip1 domain family, member 1; YIP1 family member 1; Yipf1; YIPF1-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54432 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y548 | |
| YIPF1 | |
| Recombinant protein corresponding to Human YIPF1. Recombinant protein control fragment (Product #RP-109375). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.