missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YIF1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148308
This item is not returnable.
View return policy
Description
YIF1A Polyclonal Antibody for IHC (P)
YIF1A has a possible role in transport between endoplasmic reticulum and Golgi.
Specifications
| YIF1A | |
| Polyclonal | |
| Unconjugated | |
| YIF1A | |
| 5330422J23Rik; 54TM; 54TMp; FinGER7; HYIF1P; liver protein; protein YIF1A; putative Rab5-interacting protein; putative transmembrane protein 54TMp; putative transmembrane protein; homolog of yeast Golgi membrane protein Yif1p (Yip1p-interacting factor); Yif1; YIF1A; Yif1p; Yip1 interacting factor homolog A; Yip1 interacting factor homolog A (S. cerevisiae); Yip1 interacting factor homolog A, membrane trafficking protein; YIP1-interacting factor homolog A; Yip1p-interacting factor | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| O95070 | |
| YIF1A | |
| E. coli-derived Human YIF1A fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction