missing translation for 'onlineSavingsMsg'
Learn More

YHO-13351 free base, MedChemExpress

Product Code. 16495966
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495966 100 mg 100mg
16468037 5 mg 5mg
16451527 50 mg 50mg
16415328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495966 Supplier Medchem Express Supplier No. HY12758A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YHO-13351 free base is the prodrug of YHO-13177, which is a potent and specific inhibitor of BCRP.

YHO-13351 free base is the prodrug of YHO-13177, which is a potent and specific inhibitor of BCRP.

Specifications

Chemical Name or Material YHO-13351 free base
CAS 912288-64-3
Color Orange
Purity Grade Notes Research
Molecular Formula C26H33N3O4S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (206.77 mM; Need ultrasonic)
SMILES N#C/C(C1=CC=C(OC)C(OC)=C1)=C\C2=CC=C(N3CCC(OC(CN(CC)CC)=O)CC3)S2
Molecular Weight (g/mol) 483.62
Formula Weight 483.62
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.