missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ YH 239-EE
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
YH 239-EE is a MDM2 inhibitor. Disrupts MDM2-p53 and activates p53 activity in vitro. Prevents mitotic catastrophe and promotes survival of aged muscle stem cells (muSCs). Also inhibits proliferation of AML cells in vitro.
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-[2-(tert-butylamino)-1-[N-(4-chlorobenzyl)formamido]-2-oxoethyl]-6-chloro-1H-indole-2-carboxylate |
| CAS | 1364488-67-4 |
| Quantity | 10 mg |
| Molecular Formula | C25H27Cl2N3O4 |
| Formula Weight | 504.41 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction