missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ YH 239-EE

Product Code. 18715456
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18715456

Brand: Tocris Bioscience™ 6719/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Promotes survival of muscle stem cells (MuSCs)

YH 239-EE is a MDM2 inhibitor. Disrupts MDM2-p53 and activates p53 activity in vitro. Prevents mitotic catastrophe and promotes survival of aged muscle stem cells (muSCs). Also inhibits proliferation of AML cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3-[2-(tert-butylamino)-1-[N-(4-chlorobenzyl)formamido]-2-oxoethyl]-6-chloro-1H-indole-2-carboxylate
CAS 1364488-67-4
Quantity 10 mg
Molecular Formula C25H27Cl2N3O4
Formula Weight 504.41
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.