Learn More
YES1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13977-1-AP-150UL
This item is not returnable.
View return policy
Description
The YES1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human YES1. This antibody recognizes human antigen. The YES1 antibody has been validated for the following applications: IHC, IP, ELISA analysis.
Specifications
| YES1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 7525 | |
| IgG |
| Immunoprecipitation, Immunohistochemistry, ELISA | |
| 233 μg/mL | |
| Rabbit | |
| Primary | |
| Human |