missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YES1 Polyclonal Antibody

Product Code. 13441377
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13441377 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13441377 Supplier Invitrogen™ Supplier No. PA527228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A431, Neuro 2A, C8D30, NIH-3T3, Raw264.7, PC-12, Rat2. Predicted reactivity: Mouse (99%), Zebrafish (95%), Xenopus laevis (97%), Dog (100%), Chicken (97%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

YES1 is a recombinant human protein kinase that is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YES1
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Yes1
Gene Accession No. F1LM93, P07947, Q04736
Gene Alias cellular yes-1 protein; c-Src; c-Yes; EC 2.7.10.2; HsT441; p60c-yes; p60-Src; P61-YES; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; tyrosine-protein kinase Yes; viral oncogene yes homolog; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral (v-yes) oncogene homolog 1; Yamaguchi sarcoma viral oncogene homolog 1; Yes; YES proto-oncogene 1, Src family tyrosine kinase; Yes1; YES1 proto-oncogene, Src family tyrosine kinase
Gene Symbols Yes1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human c-Yes. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22612, 24884, 7525
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.