missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yes Antibody (S07-8B3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19980-100ul
This item is not returnable.
View return policy
Description
Yes Monoclonal antibody specifically detects Yes in Human samples. It is validated for Western Blot
Specifications
| Yes | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S07-8B3 | |
| Unconjugated | |
| cellular yes-1 protein, c-yes, EC 2.7.10, EC 2.7.10.2, HsT441, P61-YES, Proto-oncogene c-Yes, proto-oncogene tyrosine-protein kinase Yes, v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1, Yamaguchi sarcoma oncogene, Yes | |
| A synthetic peptide of human Yes (Uniprot # P07947) | |
| 100 μg | |
| Protein Kinase | |
| 7525 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction