Learn More
Clark Scientific™ PVC Large Yellow Spill Trays
Manufactured from rigid PVC, these large yellow plastic spill trays are for use in radiation areas.
Brand: Clark Scientific™ YPVC Large
Code : 03
Additional Details : Weight : 0.50000kg
Description
- For use with radiation areas
- Manufactured from rigid PVC
- Available in yellow for use in radiation areas
- Available in white for use outside radiation areas
- Colourless PVC liners are available
Order Info
Size
Specifications
Yellow | |
Spill Tray Bench | |
1 Ea. |
PVC | |
Radiation areas | |
Large |