missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yellow Fever Virus NS1 Mouse anti-Virus, Clone: CE6.A.B9.G3, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Yellow Fever Virus NS1 Monoclonal antibody specifically detects Yellow Fever Virus NS1 in Virus samples. It is validated for ELISA, Immunofluorescence, ELISA, Direct ELISA
Specifications
Specifications
| Antigen | Yellow Fever Virus NS1 |
| Applications | ELISA, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Clone | CE6.A.B9.G3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA, Direct ELISA |
| Formulation | PBS pH 7.4 |
| Host Species | Mouse |
| Immunogen | Recombinant Yellow Fever virus NS1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?