Learn More
YEATS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24717-1-AP-150UL
 View more versions of this product
Produktkode 16813205
  • £349.00 / 150µL
Denne vare kan ikke returneres. Se returpolitik
Description
Description
The YEATS domain-containing protein 2 (YEATS2) is a nuclear protein that is phosphorylated upon DNA damage by ATM or ATR and contains 1 YEATS domain. The gene encoding YEATS2 resides on chromosome 3, which contains about 214 million bases encoding over 1, 100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B.
Specifications
Specifications
YEATS2
Polyclonal
Unconjugated
YEATS2
KIAA1197, YEATS domain containing 2, YEATS2
Rabbit
Antigen Affinity Chromatography
RUO
55689
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9ULM3
YEATS2
YEATS2 Fusion Protein Ag20134
150 μL