missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Yeast PST1 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Pst1 Synthetic Peptide, PEP-1456, from Invitrogen.
Protoplast secreted protein 1 (Pst1) is an anchored plasma membrane protein. Pst1 was previously identified as a protein secreted by yeast regenerating protoplasts, which suggests a role in cell wall construction. Pst1 is a protein with 444 amino acids and is attached to yeast cell wall via a glycosylphosphatidylinisotol (GPI) anchor. Pst1 contains 15 potential N-linked glycosylation sites and is heavily glycosylated. It migrates at approximately 200 kDa on SDS-PAGE when produced in wild type S. cerevisiae. Our Pst1 protein was expressed in a genetically manipulated triple-mutant (TM) S. cerevisiae stain, which results in the production of sole Man8GlcNAc2 carbohydrate structures and Pst1 migration at approximately 100 kDa. When produced in the TM yeast, Pst1 can be recognized by several glycan-specific HIV-1 broadly neutralizing antibodies, including 2G12 and recently identified PGT antibodies. Among several heavily N-glycosylated TM yeast glycoproteins, Pst1 shows higher affinity for 2G12 and efficiently inhibits gp120 interactions with 2G12 and DC-SIGN, and it also blocks 2G12-mediated neutralization of HIV-1 pseudoviruses.
Specifications
Specifications
| Gene ID (Entrez) | 851625 |
| Species | Yeast |
| Purification Method | purified |
| Common Name | Yeast PST1 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction