missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast Nitrogen Base

Product Code. 11347819 Shop All MP Biomedicals Products
Change view
Click to view available options
Yeast Nitrogen Base; Weight: 454 g; Format: Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11347819 Yeast Nitrogen Base; Weight: 454 g; Format: Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11347819 Supplier MP Biomedicals™ Supplier No. 114029622
  • ONLINE EXCLUSIVE PRICE
    £530.00
    £496.65 / 454g
    Save £33.35
    6% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MP Biologicals™ YNB with Ammonium Sulfate without Potassium Phosphate is a component of synthetic defined yeast media.

  • Yeast Nitrogen Base is available with or without ammonium sulfate.
  • YNB is composed of salts, vitamins and trace elements. Some component dropouts are available.
  • To make your own synthetic defined yeast media, start with the YNB of your choice, add a carbon source, the appropriate amino acids, and agar if necessary.
  • Bulk powder, easily weighed to make custom batch sizes.

Specifications

Concentration 0.7 g/L
Form Powder
Grade Molecular Biology Certified™
Media Type YNB
Formulation Vitamins: Biotin 0.002 mg; Calcium Pantothenate 0.4 mg; Folic Acid 0.002 mg; Inositol 2 mg: Niacin 0.4 mg; P-Aminobenzoic Acid 0.2 mg; Pyridoxine Hydrochloride 0.4 mg; Riboflavin 0.2 mg; Thiamine Hydrochloride 0.4 mg; Trace Elements: Boric Acid 0.5 mg; Copper Sulfate 0.04 mg; Potassium Iodide 0.1 mg; Ferric Chloride 0.2 mg; Manganese Sulfate 0.4 mg; Sodium Molybdate 0.2 mg; Zinc Sulfate 0.4 mg; Salts: Magnesium Sulfate 500 mg; Sodium Chloride 100 mg; Calcium Chloride 100 mg.
pH 5.0 ±0.2
Solubility Soluble in Water
Weight (Metric) 454 g
compliance-icons
Product Identifier
  • YNB w/o Ammonium Sulfate w/o Potassium Phosphate (Powder)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: calcium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.