missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast Lytic Enzyme, Arthrobacter luteus

Product Code. 15435459
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435459 1 g 1g
15445459 250 mg 250mg
15455459 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435459 Supplier Thermo Scientific Chemicals Supplier No. J63195.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrolyzes linear glucose polymers with beta-1,3-linkages | CAS: 37340-57-1

Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37340-57-1
Synonym Lyticase; Zymolyase 20T

Specifications

Color Off-white
Product Type Cell Lysis Enzyme
Content And Storage 2°C to 8°C
Form Lyophilized
For Use With (Application) Hydrolyzes linear glucose polymers with beta-1,3-linkages
Quantity 1 g
compliance-icons
Product Identifier
  • Yeast Lytic Enzyme, Arthrobacter luteus
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.