missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Yeast Extract Large Granules

Product Code. 12881660
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12881660 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12881660 Supplier Fisher BioReagents™ Supplier No. 12881660
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used for preparation of microbiological culture media.

  • Light beige
  • Average particile size: 1-2mm

Specifications

Product Type Dehydrated Culture Media
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.