missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast extract, for biochemistry

Product Code. 17201336 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Product Code. Quantity unitSize
17201336 250 g 250g
17291326 1 kg 1kg
2 options
This item is not returnable. View return policy

Product Code. 17201336

Brand: Thermo Scientific Chemicals 468552500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8013-01-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8013-01-2

Specifications

Purity or Quality Grade Biochemistry Grade
Quantity 250 g
Chemical Name or Material Yeast extract

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.