missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast Extract Agar (Dehydrated)
GREENER_CHOICE

Product Code. 12657929
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
3kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12657929 2.5 kg 3kg
11923792 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12657929 Supplier Thermo Scientific™ Supplier No. CM0019R

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Carry out plate count of microorganisms in water with this medium while adhering to ISO 1133:2014.

Carry out plate counts of microorganisms in water with Thermo Scientific™ Oxoid™ Yeast Extract Agar (Dehydrated). Yeast Extract Agar is made to the formula described by Windle Taylor1 for the plate count of microorganisms in water and is tested in accordance with ISO 1133:20142. It is a rich, nutritious medium for the recovery of a wide range of bacteria, yeast, and molds.

The usual method of counting heterotrophic bacteria in water is by the poured plate method with Yeast Extract Agar. Separate counts are made of those aerobic mesophilic organisms which form visible colonies in this medium after 24 hours incubation at 37°C and those which form colonies after three days incubation at 22°C. The two methods give different results2 bacteria, yeast, and molds.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Agar
Quantity 2.5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.