missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Yeast extract agar CM19 Oxoid
Specifications
Specifications
| Appearance | Light straw powder |
| For Use With (Application) | Plate count agar for microorganisms in water |
| pH | 7.0 to 7.4 |
| Quantity | 5 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?