missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Yeast extract
Click to view available options
Quantity:
1 kg
5kg
25 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Format | Fiber Drum |
Quantity | 25 kg |
Chemical Name or Material | Yeast Extract |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'